COM448666726 Combi-Blocks, Inc.

N-Nonanophenone

  • MDL number MFCD00048965
  • Cas Number : 6008-36-2
  • Molecular Weight : 218.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3557

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3557
Availability details for
Quantity
  • Availability
  • Add to Cart