COM448666728 Combi-Blocks, Inc.

Acetic acid (1s,2r)-2-[n-benzyl-n-(mesitylenesulfonyl)amino]-1-phenylpropyl ester

  • MDL number MFCD02093425
  • Cas Number : 240423-53-4
  • Molecular Weight : 465.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3561

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-3561
Availability details for
Quantity
  • Availability
  • Add to Cart