COM448666824 Combi-Blocks, Inc.

2,7-Diisopropylnaphthalene

  • MDL number MFCD00671522
  • Cas Number : 40458-98-8
  • Molecular Weight : 212.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-4670

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-4670
Availability details for
Quantity
  • Availability
  • Add to Cart