COM448666854 Combi-Blocks, Inc.

Diphenyliodonium-2-carboxylate monohydrate

  • MDL number MFCD00150328
  • Cas Number : 96195-89-0
  • Molecular Weight : 342.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-5003

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-5003
Availability details for
Quantity
  • Availability
  • Add to Cart