COM448666906 Combi-Blocks, Inc.

Bis(3-nitrophenyl) disulfide

  • MDL number MFCD00007246
  • Cas Number : 537-91-7
  • Molecular Weight : 308.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-5551

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-5551
Availability details for
Quantity
  • Availability
  • Add to Cart