COM448667055 Combi-Blocks, Inc.

N-Benzylquinidinium chloride

  • MDL number MFCD00198106
  • Cas Number : 77481-82-4
  • Molecular Weight : 451.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QF-7008

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QF-7008
Availability details for
Quantity
  • Availability
  • Add to Cart