COM448667347 Combi-Blocks, Inc.

4,5-Ethylenedithio-1,3-dithiol-2-thione

  • MDL number MFCD00137882
  • Cas Number : 59089-89-3
  • Molecular Weight : 224.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-0398

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-0398
Availability details for
Quantity
  • Availability
  • Add to Cart