COM448667428 Combi-Blocks, Inc.

1,2-Benzanthraquinone

  • MDL number MFCD00003596
  • Cas Number : 2498-66-0
  • Molecular Weight : 258.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-1481

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-1481
Availability details for
Quantity
  • Availability
  • Add to Cart