COM448667464 Combi-Blocks, Inc.

4-Methoxysalicylamide

  • MDL number MFCD03840504
  • Cas Number : 6745-77-3
  • Molecular Weight : 167.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-2096

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-2096
Availability details for
Quantity
  • Availability
  • Add to Cart