COM448667566 Combi-Blocks, Inc.

Potassium bisnonafluoro-1-butanesulfonimidate

  • MDL number MFCD06200794
  • Cas Number : 129135-87-1
  • Molecular Weight : 619.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-3533

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-3533
Availability details for
Quantity
  • Availability
  • Add to Cart