COM448667567 Combi-Blocks, Inc.

1-Amino-4-propylcyclohexane

  • MDL number MFCD11156904
  • Cas Number : 102653-37-2
  • Molecular Weight : 141.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-3537

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-3537
Availability details for
Quantity
  • Availability
  • Add to Cart