COM448667591 Combi-Blocks, Inc.

Tributyl orthoformate

  • MDL number MFCD00015250
  • Cas Number : 588-43-2
  • Molecular Weight : 232.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-3869

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-3869
Availability details for
Quantity
  • Availability
  • Add to Cart