COM448667607 Combi-Blocks, Inc.

N-Methylfurohydroxamic acid

  • MDL number MFCD00144939
  • Cas Number : 109531-96-6
  • Molecular Weight : 141.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-4165

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-4165
Availability details for
Quantity
  • Availability
  • Add to Cart