COM448667614 Combi-Blocks, Inc.

Nalpha-[(9h-fluoren-9-ylmethoxy)carbonyl]-l-threonine tert-butyl ester

  • MDL number MFCD08276378
  • Cas Number : 120791-76-6
  • Molecular Weight : 397.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-4304

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-4304
Availability details for
Quantity
  • Availability
  • Add to Cart