COM448667621 Combi-Blocks, Inc.

4-Acetamidophenylacetic acid ethyl ester

  • MDL number MFCD06797104
  • Cas Number : 13475-17-7
  • Molecular Weight : 221.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-4382

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-4382
Availability details for
Quantity
  • Availability
  • Add to Cart