COM448667627 Combi-Blocks, Inc.

3-Ethyl (-)-thiazolidine-3,4-dicarboxylate

  • MDL number MFCD04117921
  • Cas Number : 127657-29-8
  • Molecular Weight : 205.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-4463

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-4463
Availability details for
Quantity
  • Availability
  • Add to Cart