COM448667636 Combi-Blocks, Inc.

m-Toluidine HCl

  • MDL number MFCD00070607
  • Cas Number : 638-03-9
  • Molecular Weight : 143.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-4552

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-4552
Availability details for
Quantity
  • Availability
  • Add to Cart