COM448667641 Combi-Blocks, Inc.

P-(1-Adamantyl)toluene

  • MDL number MFCD00181038
  • Cas Number : 1459-55-8
  • Molecular Weight : 226.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-4619

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-4619
Availability details for
Quantity
  • Availability
  • Add to Cart