COM448667643 Combi-Blocks, Inc.

Diisopropyl(bromomethyl)boronate

  • MDL number MFCD01631222
  • Cas Number : 137297-49-5
  • Molecular Weight : 222.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-4639

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-4639
Availability details for
Quantity
  • Availability
  • Add to Cart