COM448667645 Combi-Blocks, Inc.

2-Propyl-4-(4-pyridylazo)phenol

  • MDL number MFCD06797155
  • Cas Number : 253124-08-2
  • Molecular Weight : 241.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-4649

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-4649
Availability details for
Quantity
  • Availability
  • Add to Cart