COM448667653 Combi-Blocks, Inc.

S-(N,N-Dimethylthiocarbamoyl)thioglycolic acid

  • MDL number MFCD00021765
  • Cas Number : 4007-01-6
  • Molecular Weight : 179.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-4698

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-4698
Availability details for
Quantity
  • Availability
  • Add to Cart