COM448667658 Combi-Blocks, Inc.

10-Methyl-9(10H)-acridone

  • MDL number MFCD00005024
  • Cas Number : 719-54-0
  • Molecular Weight : 209.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-4757

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-4757
Availability details for
Quantity
  • Availability
  • Add to Cart