COM448667659 Combi-Blocks, Inc.

2,7-Di-tert-butylnaphthalene

  • MDL number MFCD00094397
  • Cas Number : 10275-58-8
  • Molecular Weight : 240.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-4780

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-4780
Availability details for
Quantity
  • Availability
  • Add to Cart