COM448667694 Combi-Blocks, Inc.

7-Octen-1-ol

  • MDL number MFCD00798076
  • Cas Number : 13175-44-5
  • Molecular Weight : 128.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5374

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5374
Availability details for
Quantity
  • Availability
  • Add to Cart