COM448667699 Combi-Blocks, Inc.

N-(tert-Butoxycarbonyl)phosphoramidic acid diethyl ester

  • MDL number MFCD00015667
  • Cas Number : 85232-02-6
  • Molecular Weight : 253.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5551

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5551
Availability details for
Quantity
  • Availability
  • Add to Cart