COM448667700 Combi-Blocks, Inc.

Butylsuccinic anhydride

  • MDL number MFCD00598109
  • Cas Number : 2035-76-9
  • Molecular Weight : 156.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5566

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5566
Availability details for
Quantity
  • Availability
  • Add to Cart