COM448667701 Combi-Blocks, Inc.

2-Methyl-2-nitro-1,3-propanediol

  • MDL number MFCD00024812
  • Cas Number : 77-49-6
  • Molecular Weight : 135.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5579

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5579
Availability details for
Quantity
  • Availability
  • Add to Cart