COM448667705 Combi-Blocks, Inc.

Tetrabutylphosphonium tetraphenylborate

  • MDL number MFCD04038112
  • Cas Number : 29089-62-1
  • Molecular Weight : 578.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5675

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5675
Availability details for
Quantity
  • Availability
  • Add to Cart