COM448667706 Combi-Blocks, Inc.

4,4'-Di-n-octyloxyazoxybenzene

  • MDL number MFCD00059445
  • Cas Number : 25729-12-8
  • Molecular Weight : 454.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5677

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5677
Availability details for
Quantity
  • Availability
  • Add to Cart