COM448667709 Combi-Blocks, Inc.

Trans-hydrindane

  • MDL number MFCD00085358
  • Cas Number : 3296-50-2
  • Molecular Weight : 124.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5701

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5701
Availability details for
Quantity
  • Availability
  • Add to Cart