COM448667716 Combi-Blocks, Inc.

4-Heptylbenzonitrile

  • MDL number MFCD00143348
  • Cas Number : 60484-67-5
  • Molecular Weight : 201.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5767

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5767
Availability details for
Quantity
  • Availability
  • Add to Cart