COM448667720 Combi-Blocks, Inc.

2,2-Bis(4-isocyanatophenyl)hexafluoropropane

  • MDL number MFCD00142759
  • Cas Number : 10224-18-7
  • Molecular Weight : 386.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5794

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5794
Availability details for
Quantity
  • Availability
  • Add to Cart