COM448667723 Combi-Blocks, Inc.

1,3-Dimethyl-2-thiohydantoin

  • MDL number MFCD00186242
  • Cas Number : 1801-62-3
  • Molecular Weight : 144.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5813

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5813
Availability details for
Quantity
  • Availability
  • Add to Cart