COM448667725 Combi-Blocks, Inc.

2-Propylcyclohexanone

  • MDL number MFCD00019464
  • Cas Number : 94-65-5
  • Molecular Weight : 140.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5851

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5851
Availability details for
Quantity
  • Availability
  • Add to Cart