COM448667732 Combi-Blocks, Inc.

4-(5-Nonyl)pyridine

  • MDL number MFCD00010710
  • Cas Number : 2961-47-9
  • Molecular Weight : 205.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5930

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5930
Availability details for
Quantity
  • Availability
  • Add to Cart