COM448667733 Combi-Blocks, Inc.

3,5-Dibromo-1-trimethylsilylbenzene

  • MDL number MFCD03844787
  • Cas Number : 17878-23-8
  • Molecular Weight : 308.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5936

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5936
Availability details for
Quantity
  • Availability
  • Add to Cart