COM448667734 Combi-Blocks, Inc.

2,5-Dimethylfuran-3-thiol

  • MDL number MFCD00801026
  • Cas Number : 55764-23-3
  • Molecular Weight : 128.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5946

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5946
Availability details for
Quantity
  • Availability
  • Add to Cart