COM448667735 Combi-Blocks, Inc.

Cis-1,3-dimethylcyclopentane

  • MDL number MFCD00045428
  • Cas Number : 2532-58-3
  • Molecular Weight : 98.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5970

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5970
Availability details for
Quantity
  • Availability
  • Add to Cart