COM448667736 Combi-Blocks, Inc.

2-Methoxy-1-butanol

  • MDL number MFCD08276387
  • Cas Number : 15467-25-1
  • Molecular Weight : 104.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5978

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-5978
Availability details for
Quantity
  • Availability
  • Add to Cart