COM448667743 Combi-Blocks, Inc.

2-Isobutyl-4-methylthiazole

  • MDL number MFCD00053138
  • Cas Number : 61323-24-8
  • Molecular Weight : 155.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6038

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6038
Availability details for
Quantity
  • Availability
  • Add to Cart