COM448667747 Combi-Blocks, Inc.

N-Cinnamoyl-n-(2,3-xylyl)hydroxylamine

  • MDL number MFCD00059308
  • Cas Number : 69891-38-9
  • Molecular Weight : 267.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6062

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6062
Availability details for
Quantity
  • Availability
  • Add to Cart