COM448667754 Combi-Blocks, Inc.

2,6-Dimethyl-1-heptene

  • MDL number MFCD00048377
  • Cas Number : 3074-78-0
  • Molecular Weight : 126.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6136

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6136
Availability details for
Quantity
  • Availability
  • Add to Cart