COM448667767 Combi-Blocks, Inc.

2,3,4,5-Tetrachlorobenzoyl chloride

  • MDL number MFCD08460213
  • Cas Number : 42221-52-3
  • Molecular Weight : 278.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6305

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6305
Availability details for
Quantity
  • Availability
  • Add to Cart