COM448667769 Combi-Blocks, Inc.

Acetyl sulfide

  • MDL number MFCD00008753
  • Cas Number : 3232-39-1
  • Molecular Weight : 118.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6335

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6335
Availability details for
Quantity
  • Availability
  • Add to Cart