COM448667770 Combi-Blocks, Inc.

9-Acetyl-3,6-diiodocarbazole

  • MDL number MFCD10000947
  • Cas Number : 606129-89-9
  • Molecular Weight : 461.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6351

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6351
Availability details for
Quantity
  • Availability
  • Add to Cart