COM448667772 Combi-Blocks, Inc.

4-Diazo-4'-methoxydiphenylamine sulfate

  • MDL number MFCD00080910
  • Cas Number : 49732-38-9
  • Molecular Weight : 323.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6395

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6395
Availability details for
Quantity
  • Availability
  • Add to Cart