COM448667785 Combi-Blocks, Inc.

1-Chloro-2-(chloromethyl)-3,5-dioxahexane

  • MDL number MFCD00130114
  • Cas Number : 70905-45-2
  • Molecular Weight : 173.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6617

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6617
Availability details for
Quantity
  • Availability
  • Add to Cart