COM448667793 Combi-Blocks, Inc.

2,2,5-Trimethylhexane

  • MDL number MFCD00026525
  • Cas Number : 3522-94-9
  • Molecular Weight : 128.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6725

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6725
Availability details for
Quantity
  • Availability
  • Add to Cart