COM448667794 Combi-Blocks, Inc.

Allylsuccinic anhydride

  • MDL number MFCD00085227
  • Cas Number : 7539-12-0
  • Molecular Weight : 140.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6763

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6763
Availability details for
Quantity
  • Availability
  • Add to Cart