COM448667796 Combi-Blocks, Inc.

1-Ethyl-3-methylcyclohexane

  • MDL number MFCD00045510
  • Cas Number : 3728-55-0
  • Molecular Weight : 126.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6784

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QG-6784
Availability details for
Quantity
  • Availability
  • Add to Cart